Learn More
p-Phenylenediamine dihydrochloride, 99+%, Thermo Scientific Chemicals
$82.09 - $194.49
Chemical Identifiers
CAS | 624-18-0 |
---|---|
Molecular Formula | C6H8N2·2HCl |
Molecular Weight (g/mol) | 181.06 |
MDL Number | MFCD00013002 |
InChI Key | IYXMNTLBLQNMLM-UHFFFAOYSA-N |
Synonym | benzene-1,4-diamine dihydrochloride, p-phenylenediamine dihydrochloride, 1,4-diaminobenzene dihydrochloride, 1,4-phenylenediamine dihydrochloride, fourrine ds, 1,4-benzenediamine, dihydrochloride, pelagol cd, pelagol grey cd, durafur black rc, fourrine 64 |
PubChem CID | 12205 |
IUPAC Name | benzene-1,4-diamine;dihydrochloride |
SMILES | C1=CC(=CC=C1N)N.Cl.Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC417480250
|
Thermo Scientific Chemicals
417480250 |
25 g | Glass bottle |
Each for $82.09
|
|
||||
AC417481000
|
Thermo Scientific Chemicals
417481000 |
100 g | Glass bottle |
Each for $194.49
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
624-18-0 | |
181.06 | |
IYXMNTLBLQNMLM-UHFFFAOYSA-N | |
12205 | |
C1=CC(=CC=C1N)N.Cl.Cl |
C6H8N2·2HCl | |
MFCD00013002 | |
benzene-1,4-diamine dihydrochloride, p-phenylenediamine dihydrochloride, 1,4-diaminobenzene dihydrochloride, 1,4-phenylenediamine dihydrochloride, fourrine ds, 1,4-benzenediamine, dihydrochloride, pelagol cd, pelagol grey cd, durafur black rc, fourrine 64 | |
benzene-1,4-diamine;dihydrochloride |
Specifications
624-18-0 | |
Authentic | |
Glass bottle | |
C6H4(NH2)2·2HCl | |
25 g | |
Solubility in water: soluble. | |
C1=CC(=CC=C1N)N.Cl.Cl | |
181.06 | |
181.06 | |
Crystalline Powder |
Gray-Purple to White | |
99% min. (Argentometry) | |
C6H8N2·2HCl | |
MFCD00013002 | |
benzene-1,4-diamine dihydrochloride, p-phenylenediamine dihydrochloride, 1,4-diaminobenzene dihydrochloride, 1,4-phenylenediamine dihydrochloride, fourrine ds, 1,4-benzenediamine, dihydrochloride, pelagol cd, pelagol grey cd, durafur black rc, fourrine 64 | |
IYXMNTLBLQNMLM-UHFFFAOYSA-N | |
benzene-1,4-diamine;dihydrochloride | |
12205 | |
99+% | |
p-Phenylenediamine dihydrochloride, 99+% |
Safety and Handling
GHS H Statement:
Toxic if inhaled.
Toxic in contact with skin.
Very toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.
Toxic if swallowed.
Causes serious eye irritation.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Wear eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 210-834-9
RTECSNumber : ST0350000
TSCA : TSCA
RUO – Research Use Only