missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
p-Toluenesulfonic Acid, 12% in Acetic Acid, Thermo Scientific Chemicals
$273.84 - $524.96
Specifications
Chemical Name or Material | p-Toluenesulfonic acid |
---|---|
Color | Colorless to Yellow |
Name Note | 12% in Acetic Acid |
CAS | 104-15-4,64-19-7 |
Boiling Point | 116.0°C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | ||||
AC421222500
|
Thermo Scientific Chemicals
421222500 |
250 g | Glass bottle |
Each for $273.84
|
|
||||
AC421220010
|
Thermo Scientific Chemicals
421220010 |
1 kg | Glass bottle |
Each for $524.96
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
p-Toluenesulfonic acid | |
12% in Acetic Acid | |
116.0°C | |
MFCD00064387 | |
11 to 13% (Alkalimetry) | |
1.0700g/mL | |
p-toluenesulfonic acid, p-toluenesulphonic acid, 4-toluenesulfonic acid, tosic acid, p-tolylsulfonic acid, toluene-4-sulfonic acid, toluenesulfonic acid, tosylic acid, benzenesulfonic acid, 4-methyl, p-methylbenzenesulfonic acid | |
JOXIMZWYDAKGHI-UHFFFAOYSA-N | |
4-methylbenzenesulfonic acid | |
6101 | |
172.2 | |
Glass bottle | |
1.07 | |
Solution |
Colorless to Yellow | |
104-15-4,64-19-7 | |
C7H8O3S | |
11, 97 | |
01,1172; 04,508; 05,673; 06,597; 07,374; 08,488; 09,471; 11,535; 12,507 | |
15, 9692 | |
Solubility in water: soluble. | |
CC1=CC=C(C=C1)S(=O)(=O)O | |
172.198 | |
CHEBI:27849 | |
Pure | |
1.3825 to 1.3845 | |
41°C |
RUO – Research Use Only