missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Toluenesulfonic acid monohydrate, 99%, extra pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 139021000

 View more versions of this product

Catalog No. AC139021000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

p-Toluenesulfonic acid monohydrate
100.0°C to 108.0°C
99%
CH3C6H4SO3H·H2O
01,1172; 04,508; 05,673; 06,597; 07,374; 08,488; 09,471; 11,535; 12,507
Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and chloroform
CC1=CC=C(C=C1)S(=O)(=O)O.O
190.213
190.22
Extra Pure
180°C
>98.5% (total TSA)
6192-52-5
C7H10O4S
MFCD00142137
p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate
KJIFKLIQANRMOU-UHFFFAOYSA-N
4-methylbenzenesulfonic acid;hydrate
521998
99%
100 g
Plastic bottle

Chemical Identifiers

6192-52-5
190.213
KJIFKLIQANRMOU-UHFFFAOYSA-N
521998
CC1=CC=C(C=C1)S(=O)(=O)O.O
C7H10O4S
MFCD00142137
p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate
4-methylbenzenesulfonic acid;hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

RTECSNumber : DB7164000

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only