Learn More
p-Toluenesulfonic acid monohydrate, 99%, extra pure, Thermo Scientific Chemicals
$46.01 - $688.69
Chemical Identifiers
CAS | 6192-52-5 |
---|---|
Molecular Formula | C7H10O4S |
Molecular Weight (g/mol) | 190.213 |
MDL Number | MFCD00142137 |
InChI Key | KJIFKLIQANRMOU-UHFFFAOYSA-N |
Synonym | p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate |
PubChem CID | 521998 |
IUPAC Name | 4-methylbenzenesulfonic acid;hydrate |
SMILES | CC1=CC=C(C=C1)S(=O)(=O)O.O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC139020050
|
Thermo Scientific Chemicals
139020050 |
5 g | Glass Bottle |
Each for $46.01
|
|
|||||
AC139021000
|
Thermo Scientific Chemicals
139021000 |
100 g | Plastic bottle |
Each for $74.40
|
|
|||||
AC139025000
|
Thermo Scientific Chemicals
139025000 |
500 g | Plastic bottle |
Each for $155.41
|
|
|||||
AC139020025
|
Thermo Scientific Chemicals
139020025 |
2.5 kg | Plastic bottle |
Each for $224.50
|
|
|||||
AC139020100
|
Thermo Scientific Chemicals
139020100 |
10 kg | Plastic drum |
Each for $688.69
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6192-52-5 | |
190.213 | |
KJIFKLIQANRMOU-UHFFFAOYSA-N | |
521998 | |
CC1=CC=C(C=C1)S(=O)(=O)O.O |
C7H10O4S | |
MFCD00142137 | |
p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate | |
4-methylbenzenesulfonic acid;hydrate |
Specifications
100.0°C to 108.0°C | |
99% | |
CH3C6H4SO3H·H2O | |
01,1172; 04,508; 05,673; 06,597; 07,374; 08,488; 09,471; 11,535; 12,507 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and chloroform | |
CC1=CC=C(C=C1)S(=O)(=O)O.O | |
190.213 | |
190.22 | |
Extra Pure | |
180°C | |
p-Toluenesulfonic acid monohydrate, >98.5% (total TSA) |
6192-52-5 | |
C7H10O4S | |
MFCD00142137 | |
p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate | |
KJIFKLIQANRMOU-UHFFFAOYSA-N | |
4-methylbenzenesulfonic acid;hydrate | |
521998 | |
99% | |
5 g | |
Glass Bottle |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RTECSNumber : DB7164000
RUO – Research Use Only