Learn More
p-Toluenesulfonylacetic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0210206
Description
p-Toluenesulfonylacetic acid is used as pharmaceuticals intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsp-Toluenesulfonylacetic acid is used as pharmaceuticals intermediates.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
p-Toluenesulfonylacetic acid | |
117°C to 119°C | |
MFCD00021764 | |
2331311 | |
Insoluble in water. | |
CC1=CC=C(C=C1)S(=O)(=O)CC(=O)O | |
214.235 | |
214.24 |
3937-96-0 | |
C9H10O4S | |
5 g | |
2-tosylacetic acid, 4-toluenesulfonylacetic acid, 4-methylphenyl sulfonyl acetic acid, 2-p-toluenesulfonyl acetic acid, tosylacetic acid, unii-yid78nf77o, p-toluenesulphonylacetic acid, 4-toluenesulphonylacetic acid, p-toluenesulfonylacetic acid, yid78nf77o | |
AQDHXMBUTDLAMD-UHFFFAOYSA-N | |
2-(4-methylphenyl)sulfonylacetic acid | |
77538 | |
98% |
Chemical Identifiers
3937-96-0 | |
214.235 | |
AQDHXMBUTDLAMD-UHFFFAOYSA-N | |
77538 | |
CC1=CC=C(C=C1)S(=O)(=O)CC(=O)O |
C9H10O4S | |
MFCD00021764 | |
2-tosylacetic acid, 4-toluenesulfonylacetic acid, 4-methylphenyl sulfonyl acetic acid, 2-p-toluenesulfonyl acetic acid, tosylacetic acid, unii-yid78nf77o, p-toluenesulphonylacetic acid, 4-toluenesulphonylacetic acid, p-toluenesulfonylacetic acid, yid78nf77o | |
2-(4-methylphenyl)sulfonylacetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 223-518-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only