missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Toluenesulfonylacetic acid, 98%, Thermo Scientific Chemicals

$177.75 - $748.22

Chemical Identifiers

CAS 3937-96-0
Molecular Formula C9H10O4S
Molecular Weight (g/mol) 214.235
MDL Number MFCD00021764
InChI Key AQDHXMBUTDLAMD-UHFFFAOYSA-N
Synonym 2-tosylacetic acid, 4-toluenesulfonylacetic acid, 4-methylphenyl sulfonyl acetic acid, 2-p-toluenesulfonyl acetic acid, tosylacetic acid, unii-yid78nf77o, p-toluenesulphonylacetic acid, 4-toluenesulphonylacetic acid, p-toluenesulfonylacetic acid, yid78nf77o
PubChem CID 77538
IUPAC Name 2-(4-methylphenyl)sulfonylacetic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)CC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0210206
View Documents
Thermo Scientific Chemicals
L0210206
5 g
Each for $177.75
 
AAL0210214
View Documents
Thermo Scientific Chemicals
L0210214
25 g
Each for $748.22
 
Description

Description

p-Toluenesulfonylacetic acid is used as pharmaceuticals intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Toluenesulfonylacetic acid is used as pharmaceuticals intermediates.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

3937-96-0
214.235
AQDHXMBUTDLAMD-UHFFFAOYSA-N
77538
CC1=CC=C(C=C1)S(=O)(=O)CC(=O)O
C9H10O4S
MFCD00021764
2-tosylacetic acid, 4-toluenesulfonylacetic acid, 4-methylphenyl sulfonyl acetic acid, 2-p-toluenesulfonyl acetic acid, tosylacetic acid, unii-yid78nf77o, p-toluenesulphonylacetic acid, 4-toluenesulphonylacetic acid, p-toluenesulfonylacetic acid, yid78nf77o
2-(4-methylphenyl)sulfonylacetic acid

Specifications

3937-96-0
C9H10O4S
5 g
2-tosylacetic acid, 4-toluenesulfonylacetic acid, 4-methylphenyl sulfonyl acetic acid, 2-p-toluenesulfonyl acetic acid, tosylacetic acid, unii-yid78nf77o, p-toluenesulphonylacetic acid, 4-toluenesulphonylacetic acid, p-toluenesulfonylacetic acid, yid78nf77o
AQDHXMBUTDLAMD-UHFFFAOYSA-N
2-(4-methylphenyl)sulfonylacetic acid
77538
98%
117°C to 119°C
MFCD00021764
2331311
Insoluble in water.
CC1=CC=C(C=C1)S(=O)(=O)CC(=O)O
214.235
214.24
p-Toluenesulfonylacetic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 223-518-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only