missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Toluenesulfonylacetic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0210214

 View more versions of this product

Catalog No. AAL0210214


Explore available promotions

Description

Description

p-Toluenesulfonylacetic acid is used as pharmaceuticals intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Toluenesulfonylacetic acid is used as pharmaceuticals intermediates.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

p-Toluenesulfonylacetic acid
117°C to 119°C
MFCD00021764
2331311
Insoluble in water.
CC1=CC=C(C=C1)S(=O)(=O)CC(=O)O
214.235
214.24
3937-96-0
C9H10O4S
25 g
2-tosylacetic acid, 4-toluenesulfonylacetic acid, 4-methylphenyl sulfonyl acetic acid, 2-p-toluenesulfonyl acetic acid, tosylacetic acid, unii-yid78nf77o, p-toluenesulphonylacetic acid, 4-toluenesulphonylacetic acid, p-toluenesulfonylacetic acid, yid78nf77o
AQDHXMBUTDLAMD-UHFFFAOYSA-N
2-(4-methylphenyl)sulfonylacetic acid
77538
98%

Chemical Identifiers

3937-96-0
214.235
AQDHXMBUTDLAMD-UHFFFAOYSA-N
77538
CC1=CC=C(C=C1)S(=O)(=O)CC(=O)O
C9H10O4S
MFCD00021764
2-tosylacetic acid, 4-toluenesulfonylacetic acid, 4-methylphenyl sulfonyl acetic acid, 2-p-toluenesulfonyl acetic acid, tosylacetic acid, unii-yid78nf77o, p-toluenesulphonylacetic acid, 4-toluenesulphonylacetic acid, p-toluenesulfonylacetic acid, yid78nf77o
2-(4-methylphenyl)sulfonylacetic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 223-518-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only