missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
p-Xylene-2-sulfonic Acid Hydrate 98.0+%, TCI America™
$61.38 - $238.89
Chemical Identifiers
CAS | 609-54-1 |
---|---|
Molecular Formula | C8H10O3S |
Molecular Weight (g/mol) | 186.23 |
MDL Number | MFCD00151010 |
InChI Key | IRLYGRLEBKCYPY-UHFFFAOYSA-N |
Synonym | p-xylene-2-sulfonic acid, unii-l1sea5qnp9, p-xylene-2-sulphonic acid, benzenesulfonic acid, 2,5-dimethyl, l1sea5qnp9, 2,4-xylenesulfonic acid hydrate, 2,5-dimethyl-benzenesulfonic acid, 2,5-xylenesulfonic acid, p-xylenesulfonic acid, dsstox_cid_21641 |
PubChem CID | 11868 |
IUPAC Name | 2,5-dimethylbenzene-1-sulfonic acid |
SMILES | CC1=CC=C(C)C(=C1)S(O)(=O)=O |
Chemical Identifiers
609-54-1 | |
186.23 | |
IRLYGRLEBKCYPY-UHFFFAOYSA-N | |
11868 | |
CC1=CC=C(C)C(=C1)S(O)(=O)=O |
C8H10O3S | |
MFCD00151010 | |
p-xylene-2-sulfonic acid, unii-l1sea5qnp9, p-xylene-2-sulphonic acid, benzenesulfonic acid, 2,5-dimethyl, l1sea5qnp9, 2,4-xylenesulfonic acid hydrate, 2,5-dimethyl-benzenesulfonic acid, 2,5-xylenesulfonic acid, p-xylenesulfonic acid, dsstox_cid_21641 | |
2,5-dimethylbenzene-1-sulfonic acid |
Specifications
86°C | |
Red-White | |
Crystal-Powder at 20°C | |
MFCD00151010 | |
IRLYGRLEBKCYPY-UHFFFAOYSA-N | |
2,5-dimethylbenzene-1-sulfonic acid | |
11868 | |
≥98.0% (HPLC,T) |
609-54-1 | |
25 g | |
C8H10O3S | |
p-xylene-2-sulfonic acid, unii-l1sea5qnp9, p-xylene-2-sulphonic acid, benzenesulfonic acid, 2,5-dimethyl, l1sea5qnp9, 2,4-xylenesulfonic acid hydrate, 2,5-dimethyl-benzenesulfonic acid, 2,5-xylenesulfonic acid, p-xylenesulfonic acid, dsstox_cid_21641 | |
CC1=CC=C(C)C(=C1)S(O)(=O)=O | |
186.23 | |
186.23 | |
p-Xylene-2-sulfonic Acid Hydrate |
Safety and Handling
EINECSNumber : (9)-2093
TSCA : Yes