Motic™ PA53 BIO LUMOS EPI fluorescence LED Trinocular Upright Research Microscope
The PA53 BIO FS6 model offers maximum flexibility for managing any kind of fluorescence application thanks to its LUMOS High-power LED FL light source, with individual control of each LED channel.
Supplier: Motic™ 1100104600931
Description
- Scalable Infinity-Corrected Optical System (CCIS™)
- Full power transmitted Köhler illumination
- Coded fluorescence turret automatically remembers fluorescence channel if changed
- LUMOS FL is great light source for fluorescence
- Unlike traditional mercury-arc lamps which take up to 20 minutes to reach constant brightness, the LUMOS FL is ready for use at the flip of a switch
- Individually control each LED channel
- Motic™ Fluorescence Imaging Software plug-in
Specifications
LUMOS Flourescence LED | |
UC Fluor Plan 4X, 10X, 20X, 40X, 100X | |
Transmitted Halogen |
10X | |
Trinocular | |
Widefield (FN23) 10X/23 mm |