Motic™ PA53 BIO LUMOS EPI fluorescence LED Trinocular Upright Research Microscope
The PA53 BIO FS6 model offers maximum flexibility for managing any kind of fluorescence application thanks to its LUMOS High-power LED FL light source, with individual control of each LED channel.
Supplier:  Motic™ 1100104601151
 View more versions of this product
Catalog No. 01-257-422
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Scalable Infinity-Corrected Optical System (CCIS™)
  • Full power transmitted Köhler illumination
  • Coded fluorescence turret automatically remembers fluorescence channel if changed
  • LUMOS FL is great light source for fluorescence
  • Unlike traditional mercury-arc lamps which take up to 20 minutes to reach constant brightness, the LUMOS FL is ready for use at the flip of a switch
  • Individually control each LED channel
  • Motic™ Fluorescence Imaging Software plug-in
Specifications
Specifications
3D-EDF LUMOS Flourescence LED
UC Fluor Plan 4X, 10X, 20X, 40X, 100X
Transmitted Halogen
10X
Trinocular
Widefield (FN23) 10X/23 mm