Motic™ PA53 MET Trinocular Upright Materials Microscope
A combination of technology and creativity, the PA53 MET series offers superior optical performance as well as an advanced ergonomic design, futuristic digital capabilities, and innovative modules ideal for metallurgy.
Supplier: Motic™ 1100104600891
Description
- Scalable Infinity-Corrected Optical System (CCIS™)
- Superior optical performance with newly designed Semi-Apochromatic objectives
- Interference Contrast (DIC) concept incident illumination
- Established contrast methods
- Multiple degrees of motorization in X, Y, and Z
- Harmonized Brightfield/Darkfield switch
- Transmitted Light: LB daylight filter (Halogen lamp)
- Diopter adjustable
Specifications
10X | |
PA53 MET BD | |
Reflected and Transmitted Illumination |
S-APO BD Plan 5X, 10X, 20X, 50X | |
Trinocular | |
WF 10X/25 mm |