Motic™ PA53 MET Trinocular Upright Materials Microscope
A combination of technology and creativity, the PA53 MET series offers superior optical performance as well as an advanced ergonomic design, futuristic digital capabilities, and innovative modules ideal for metallurgy.
$26829.26 - $31931.19
Catalog Number | Mfr. No. | Lighting Type | Model | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Lighting Type | Model | Price | Quantity | ||||
01-257-428
|
Motic™
1100104600881 |
Reflected Illumination | PA53 MET BD |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
01-257-429
|
Motic™
1100104600861 |
Reflected and Transmitted Illumination | PA53 MET |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
01-257-430
|
Motic™
1100104600891 |
Reflected and Transmitted Illumination | PA53 MET BD |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Scalable Infinity-Corrected Optical System (CCIS™)
- Superior optical performance with newly designed Semi-Apochromatic objectives
- Interference Contrast (DIC) concept incident illumination
- Established contrast methods
- Multiple degrees of motorization in X, Y, and Z
- Harmonized Brightfield/Darkfield switch
- Transmitted Light: LB daylight filter (Halogen lamp)
- Diopter adjustable
Specifications
10X | |
Trinocular | |
WF 10X/25 mm |