missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Pacific Hemostasis™ Prothrombin Time (PT) Reagents

Detect acquired or congenital disorders affecting the extrinsic and common pathways.

Supplier:  Thermo Scientific™ 100357

 View more versions of this product

Catalog No. 17-609-9

Explore available promotions

Description

Description

Detect acquired or congenital disorders affecting the extrinsic and common pathways with Thermo Scientific™ Pacific Hemostasis™ Prothrombin Time (PT) Reagents. Widely accepted as a means of monitoring patients on oral anticoagulant therapy such as Warfarin/Coumadin, reagents are manufactured with up to three years extended shelf life and have excellent lot-to-lot consistency.

Features:
  • Compatible with automated, semi-automated or manual systems
  • Reconstitution stability: 7 days at 2° to 8°C (36° to 46°F)
  • Dried rabbit brain thromboplastin

Thromboplastin-D with CaCl2 (Product No. 10-0356, 10-0357):

  • Target ISI value of 1.7 to 2.2
  • Expected normal values of 12 to 15 seconds

Thromboplastin-DS (Product No. 10-0362, 10-0354):

  • Target ISI value of 1.0 to 1.3
  • Expected normal values of 12 to 15 seconds
Specifications

Specifications

Thromboplastin-D
Prothrombin Time (PT)
10 x 10 mL
Videos
SDS
Documents

Documents

Promotions

Promotions