missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Pacific Hemostasis™ Prothrombin Time (PT) Reagents

Detect acquired or congenital disorders affecting the extrinsic and common pathways.

$114.24 - $247.32

Specifications

Type Prothrombin Time (PT)
View More Specs

Products 4
Catalog Number Mfr. No. Description Quantity Price Quantity  
Catalog Number Mfr. No. Description Quantity Price Quantity  
29-227-3
View Documents
Thermo Scientific™
100354
Thromboplastin-DS 10 x 4 mL
Pack of 10 Each for $140.79
 
29-227-4
View Documents
Thermo Scientific™
100362
Thromboplastin-DS 10 x 10 mL
Pack of 10 Each for $247.32
 
CMS-176-065
View Documents
Thermo Scientific™
100356
Thromboplastin-D 10 x 4 mL
Each for $114.24
 
17-609-9
View Documents
Thermo Scientific™
100357
Thromboplastin-D 10 x 10 mL
Pack of 500 Each for $162.41
 
Description

Description

Detect acquired or congenital disorders affecting the extrinsic and common pathways with Thermo Scientific™ Pacific Hemostasis™ Prothrombin Time (PT) Reagents. Widely accepted as a means of monitoring patients on oral anticoagulant therapy such as Warfarin/Coumadin, reagents are manufactured with up to three years extended shelf life and have excellent lot-to-lot consistency.

Features:
  • Compatible with automated, semi-automated or manual systems
  • Reconstitution stability: 7 days at 2° to 8°C (36° to 46°F)
  • Dried rabbit brain thromboplastin

Thromboplastin-D with CaCl2 (Product No. 10-0356, 10-0357):

  • Target ISI value of 1.7 to 2.2
  • Expected normal values of 12 to 15 seconds

Thromboplastin-DS (Product No. 10-0362, 10-0354):

  • Target ISI value of 1.0 to 1.3
  • Expected normal values of 12 to 15 seconds
Specifications

Specifications

Prothrombin Time (PT)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'