missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Palladium(II) acetate, 47.5% Pd, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 195180020

 View more versions of this product

Catalog No. AC195180020


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Palladium(II) acetate
47.5% Pd
2 g
(CH3CO2)2Pd
15,7092
Solubility in water: insoluble. Other solubilities: soluble in acetone, benzene, methylene dichloride,, chloroform, acetonitrile and diethylether, insoluble in alcohols
[Pd++].CC([O-])=O.CC([O-])=O
224.51
224.51
Glass bottle
205.0°C
3375-31-3
C4H6O4Pd
MFCD00012453
palladium ii acetate, palladium diacetate, palladium acetate, diacetoxypalladium, bis acetato palladium, bisacetylpalladium, diacetatopalladium, palladium 2+ acetate, palladous acetate
YJVFFLUZDVXJQI-UHFFFAOYSA-L
palladium(2+) diacetate
167845
47.5% (Pd)

Chemical Identifiers

3375-31-3
224.51
YJVFFLUZDVXJQI-UHFFFAOYSA-L
167845
C4H6O4Pd
MFCD00012453
palladium ii acetate, palladium diacetate, palladium acetate, diacetoxypalladium, bis acetato palladium, bisacetylpalladium, diacetatopalladium, palladium 2+ acetate, palladous acetate
[Pd++].CC([O-])=O.CC([O-])=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
May cause long lasting harmful effects to aquatic life.

GHS P Statement
Wear eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 222-164-4

RTECSNumber : AJ1900000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only