Learn More
Palladium(II) acetate, 47.5% Pd, Thermo Scientific Chemicals
$676.25 - $676.25
Chemical Identifiers
CAS | 3375-31-3 |
---|---|
Molecular Formula | C4H6O4Pd |
Molecular Weight (g/mol) | 224.51 |
MDL Number | MFCD00012453 |
InChI Key | YJVFFLUZDVXJQI-UHFFFAOYSA-L |
Synonym | palladium ii acetate, palladium diacetate, palladium acetate, diacetoxypalladium, bis acetato palladium, bisacetylpalladium, diacetatopalladium, palladium 2+ acetate, palladous acetate |
PubChem CID | 167845 |
SMILES | [Pd++].CC([O-])=O.CC([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC195180020
|
Thermo Scientific Chemicals
195180020 |
2 g | Glass bottle |
Each for $676.25
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3375-31-3 | |
224.51 | |
YJVFFLUZDVXJQI-UHFFFAOYSA-L | |
167845 |
C4H6O4Pd | |
MFCD00012453 | |
palladium ii acetate, palladium diacetate, palladium acetate, diacetoxypalladium, bis acetato palladium, bisacetylpalladium, diacetatopalladium, palladium 2+ acetate, palladous acetate | |
[Pd++].CC([O-])=O.CC([O-])=O |
Specifications
205.0°C | |
2 g | |
(CH3CO2)2Pd | |
15,7092 | |
Solubility in water: insoluble. Other solubilities: soluble in acetone, benzene, methylene dichloride,, chloroform, acetonitrile and diethylether, insoluble in alcohols | |
[Pd++].CC([O-])=O.CC([O-])=O | |
224.51 | |
224.51 | |
Glass bottle |
3375-31-3 | |
C4H6O4Pd | |
MFCD00012453 | |
palladium ii acetate, palladium diacetate, palladium acetate, diacetoxypalladium, bis acetato palladium, bisacetylpalladium, diacetatopalladium, palladium 2+ acetate, palladous acetate | |
YJVFFLUZDVXJQI-UHFFFAOYSA-L | |
palladium(2+) diacetate | |
167845 | |
47.5% (Pd) | |
Palladium(II) acetate, 47.5% Pd |
Safety and Handling
GHS H Statement
Causes serious eye damage.
May cause long lasting harmful effects to aquatic life.
GHS P Statement
Wear eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 222-164-4
RTECSNumber : AJ1900000
TSCA : TSCA
RUO – Research Use Only