Parr™ 6100 Compensated Calorimeter
For measuring heats of combustion of solid or liquid fuels, waste materials, and other combustible samples.
Supplier:  Parr™ 6100CLEA
 View more versions of this product
Catalog No. 04-731-181
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Includes: Compact static-jacket calorimeter module with built-in calorimeter controller, oxygen bomb, oval bucket, accessories for installation, 6008P maintenance kit, USB port for balance and printer connection
Description
Description
  • Compact, static-jacket calorimeter
  • Operates at approximately room temperature
  • Internal microprocessor applies heat leak corrections in real time
  • Clear LCD touchscreen display
  • Removable 1108P bomb and bucket
  • Semi-automatic system for charging the bomb with oxygen
  • Conducts 4 to 7 tests per hour
  • Operator time per test approximately 6 minutes
  • 0.2% Precision Class Instrument
  • FlashROM memory and ethernet network communications
Unitt should be operated in a temperature-controlled room.
Order Info
Accessory bombs, bucket, parts and related hardware and calorimeter standards are available. For more information, contact your Fisher Scientific™ Customer Service Representative.
Specifications
Specifications
Compensated Calorimeter
Ethernet or USB
40 cm
Measuring heats of combustion of solid or liquid fuels, waste materials and other combustible samples
43 cm
Stores 1000 tests
Manual
52 to 12,000 calories
4 to 7 Tests/hr.
23 in.
ISO 9001:2008, CSA, CE, ASTM
16 in.
LCD
17 in.
Compact static-jacket calorimeter module with built-in calorimeter controller, 1108PCL oxygen bomb for chlorine service, oval bucket, accessories for installation, 6008P maintenance kit, USB port for balance and printer connection
6 min./Test
0.1 to 0.2%
0.0001°C
115 V
57 cm
Product Suggestions
Product Suggestions