Parr™ Oxygen Combustional Vessels For Calorimeter
Oxygen Combustion Bomb for 6100/6200 Calorimeters
Supplier:  Parr™ 1108
 View more versions of this product
Catalog No. 04-392-1
Explore available promotions
Includes: Alloy head and cylinder
Description
Description
  • Made of Columbium Stabilized Stainless-steel
  • 342mL
Specifications
Specifications
Oxygen Combustional Vessel
Parr 6100/6200 Calorimeter
Columbium-stabilized Stainless Steel
For determining sulfur, halogens and other elements in a broad range of combustible materials
Alloy head and cylinder