missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Parr™ Oxygen Combustional Vessels For Calorimeter
Oxygen Combustion Bomb for 6100/6200 Calorimeters
$5285.28 - $8288.28
Specifications
Product Type | Oxygen Combustional Vessel |
---|---|
For Use With (Equipment) | Parr 6100/6200 Calorimeter |
Includes | Alloy head and cylinder |
Material | Columbium-stabilized Stainless Steel |
Includes: Alloy head and cylinder
Catalog Number | Mfr. No. | For Use With (Application) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Application) | Price | Quantity | |||||
04-350-7
|
Parr™
1108CL |
Used to test waste materials and combustible solvents |
Each for $8,288.28
|
|
|||||
04-392-1
|
Parr™
1108 |
For determining sulfur, halogens and other elements in a broad range of combustible materials |
Each for $5,285.28
|
|
|||||
Description
- Made of Columbium Stabilized Stainless-steel
- 342mL
Specifications
Oxygen Combustional Vessel | |
Parr 6100/6200 Calorimeter | |
Alloy head and cylinder | |
Columbium-stabilized Stainless Steel |