missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Parter Medical Products™ Sterile Specimen Containers

Incorporate the proprietary PMP™ Sure-Seal™ integrated sealing system and the proprietary PMP Sure-Lok™ proof-of-closure system

Supplier:  Parter Medical Products™ 243017

 View more versions of this product

Catalog No. 22-150-327


Explore available promotions

Description

Description

  • Straight-sided container format featuring top and bottom positive grip knurls for ease of opening and closing when wearing protective or latex gloves
  • Stackable, leak-resistant and graduated unless otherwise noted
  • Sterile containers labeled with a security tab label and sterilized by gamma irradiation to SAL 10-3; certification of sterility available upon request
  • Manufactured from virgin polypropylene (container) and virgin polyethylene (closure) without the use of plasticizers or demolding additives; all material used is free from latex and bovine contamination
  • Manufactured under strict process control and lot numbers are fully traceable
  • 95kpa compliant; testing results available upon request
  • Rigorous testing at varying pressures without seal failure proves this container ideal for use in pneumatic tube systems and harsh transport conditions; leak performance data is available upon request
  • Available in individually wrapped (Flo-Pac™) formats
  • Closures secure at 10 to 12 in.-lb. closing force
  • Reagent or fixative pre-fills may require 15 to 17 in.-lb. closing force to prevent post-production cap-back-off
  • Standard cap colors are natural, orange and blue

Specifications

Specifications

Yes
Sterile
90 mL
Polyethylene
Tamper-evident Label
Polypropylene (Clarified)
3.0 oz.
Natural
53 mm
100/Cs.
SDS
Documents

Documents

Promotions

Promotions