missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Parter Medical Products™ Sterile Specimen Containers
Incorporate the proprietary PMP™ Sure-Seal™ integrated sealing system and the proprietary PMP Sure-Lok™ proof-of-closure system
$218.12 - $330.12
Specifications
Graduated | Yes |
---|---|
Material | Polypropylene (Clarified) |
Sterility | Sterile |
Closure Material | Polyethylene |
Closure Size | 53 mm |
Catalog Number | Mfr. No. | Capacity (English) | Capacity (Metric) | Closure Color | No. per Case | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (English) | Capacity (Metric) | Closure Color | No. per Case | Price | Quantity | ||
22-150-327
|
Parter Medical Products™
243017 |
3.0 oz. | 90 mL | Natural | 100/Cs. |
Case of 100 Each for $218.12
|
|
||
22-150-328
|
Parter Medical Products™
243516 |
3.0 oz. | 90 mL | Blue | 400/Cs. |
Case of 400 Each for $309.97
|
|
||
22-150-329
|
Parter Medical Products™
243916 |
3.0 oz. | 90 mL | Orange | 400/Cs. |
Case of 400 Each for $330.12
|
|
||
Description
- Straight-sided container format featuring top and bottom positive grip knurls for ease of opening and closing when wearing protective or latex gloves
- Stackable, leak-resistant and graduated unless otherwise noted
- Sterile containers labeled with a security tab label and sterilized by gamma irradiation to SAL 10-3; certification of sterility available upon request
- Manufactured from virgin polypropylene (container) and virgin polyethylene (closure) without the use of plasticizers or demolding additives; all material used is free from latex and bovine contamination
- Manufactured under strict process control and lot numbers are fully traceable
- 95kpa compliant; testing results available upon request
- Rigorous testing at varying pressures without seal failure proves this container ideal for use in pneumatic tube systems and harsh transport conditions; leak performance data is available upon request
- Available in individually wrapped (Flo-Pac™) formats
- Closures secure at 10 to 12 in.-lb. closing force
- Reagent or fixative pre-fills may require 15 to 17 in.-lb. closing force to prevent post-production cap-back-off
- Standard cap colors are natural, orange and blue
Specifications
Yes | |
Sterile | |
53 mm |
Polypropylene (Clarified) | |
Polyethylene | |
Tamper-evident Label |