missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Greiner Bio-One Pasteur Pipets
For use in rapid liquid transfer
Supplier: Greiner Bio-One 700361
Specifications
LDPE | |
Liquid handling | |
25/Bag, 1000/Cs. | |
0.1 mL |
Sterile | |
6.02 in. | |
Translucent | |
153 mm |