MP Biomedicals™ Phosphate Buffered Saline (PBS) Tablets
Phosphate Buffered Saline Tablets (PBS) are a balanced salt mixture for easy PBS buffer preparation for a variety of applications, including reagent preparation, diluting cells for flow cytometry, and cell culture reagent.
Supplier:  MP Biomedicals™ 092810306
 View more versions of this product
Catalog No. ICN2810306
Explore available promotions
Description
Description
  • No need for weighing and mixing chemicals
  • Simply dissolve PBS tablets in water
  • One tablet dissolved in 100 mL water yields buffer containing 0.01M phosphate, 0.0027M KCl, and 0.137M NaCl, pH 7.4 at 25°C
  • Research use only
Specifications
Specifications
No Calcium, No Magnesium
200 Tablets
MFCD00011360
[Cl-].[K+]
74.55
Phosphate Buffered Saline (PBS)
ClK
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride
Tablet