missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MP Biomedicals™ Phosphate Buffered Saline (PBS) Tablets

Phosphate Buffered Saline Tablets (PBS) are a balanced salt mixture for easy PBS buffer preparation for a variety of applications, including reagent preparation, diluting cells for flow cytometry, and cell culture reagent.

$186.26 - $604.94

Specifications

Additive No Calcium, No Magnesium
Product Type Phosphate Buffered Saline (PBS)
Form Tablet
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN2810305
View Documents
MP Biomedicals™
092810305
100 Tablets
Pack of 100 Each for $186.26
 
ICN2810306
View Documents
MP Biomedicals™
092810306
200 Tablets
Pack for $484.00
 
ICN2810307
View Documents
MP Biomedicals™
092810307
500 Tablets
Pack for $604.94
 
Description

Description

  • No need for weighing and mixing chemicals
  • Simply dissolve PBS tablets in water
  • One tablet dissolved in 100 mL water yields buffer containing 0.01M phosphate, 0.0027M KCl, and 0.137M NaCl, pH 7.4 at 25°C
  • Research use only
Specifications

Specifications

No Calcium, No Magnesium
Phosphate Buffered Saline (PBS)
Tablet
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'