missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ PCC-54™ Detergent Concentrate

A laboratory cleaning solution for labware utensils, surface materials and glassware, including beakers, pipets, lenses, mirrors; RBS-35 alternative.

$233.33 - $679.99

Specifications

Form Liquid
Product Line PCC-54™
Composition PCC-PFree, 2%- 20% Solution
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C.
Description PCC-54 Detergent Concentrate
View More Specs

Products 2
Catalog Number Mfr. No. Volume (Metric) Quantity Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Quantity Price Quantity  
PI72288
View Documents
Thermo Scientific™
72288
3 L 3 L
Pack for $233.33
 
PI72289
View Documents
Thermo Scientific™
72289
3 L 4 Pk.
Pack of 4 Each for $679.99
 
Description

Description

Thermo Scientific™ PCC-54 Detergent Concentrate is a general cleaning agent for use on many laboratory surface materials and glassware, including lenses, mirrors, and reflectors.

Highlights:

  • Removal of blood residue from glass or plastic beakers and flasks Colored dyes from porcelain mortar and pestles HIV virus from glassware, tabletops and flooring For removing radioisotopes on contaminated tabletops
  • Rinses clean, providing film-free surfaces Safe to handle, and PCC-Pfree does not contain phosphates that can harm the environment Use a 2%- 20% solution at 50°C for soaking stubborn biological residues away
Specifications

Specifications

Liquid
PCC-PFree, 2%- 20% Solution
PCC-54 Detergent Concentrate
PCC-54™
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25°C.
3 L
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.