missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3M™ PELTOR™ Optime™ 98 Ear Muffs

Optime™ 98 is versatile ear muff featuring a stainless steel headband and is recommended for TWA noise exposures up to 98 dBA.

Supplier:  3M™ 7000009670

 View more versions of this product

Catalog No. 17-388-2A



Explore available promotions

Description

Description

  • Recommended for TWA noise exposures up to 98 dBA
  • Stainless steel headband distributes weight for low pressure fit
  • Ear cup pivot points tilt for comfort that adjusts to individual wearers
  • Ear cups with liquid and foam filled cushions offer comfort for long wear and optimum seal
  • Color-coding for compliance sighting

Compliance

3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

Specifications

Specifications

Yellow
Liquid-foam
Black
Assembly, cleaning, demolition, facility maintenance, grinding, machine operations, painting, sanding, welding
H9A
25 dB
Black
ABS
Yes
Class A
Stainless Steel
Yes
1 Ea.
PELTOR™ Optime™
No
Over-the-head
Videos
SDS
Documents

Documents

Promotions

Promotions