missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3M™ PELTOR™ Optime™ 98 Ear Muffs

Optime™ 98 is versatile ear muff featuring a stainless steel headband and is recommended for TWA noise exposures up to 98 dBA.

Supplier:  3M™ 7000002326

 View more versions of this product

Catalog No. 17-388-2B



Explore available promotions

Description

Description

  • Recommended for TWA noise exposures up to 98 dBA
  • Stainless steel headband distributes weight for low pressure fit
  • Ear cup pivot points tilt for comfort that adjusts to individual wearers
  • Ear cups with liquid and foam filled cushions offer comfort for long wear and optimum seal
  • Color-coding for compliance sighting

Compliance

3M™ ear muffs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

Specifications

Specifications

Yellow
Foam
Black
Assembly, cleaning, demolition, facility maintenance, grinding, machine operations, painting, sanding, welding
H9P3E
23 dB
No
Hard Hat
Yes
Class A
Most helmets, eyewear and other safety equipment
Yes
1 Ea.
PELTOR™ Optime™
ABS
Videos
SDS
Documents

Documents

Promotions

Promotions