missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pentacene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 129790010

 View more versions of this product

Catalog No. AC129790010

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Pentacene
135-48-8
Black or Blue-Purple
97.5% min
C22H14
1 g
benzo b naphthacene, lin-dibenzanthracene, lin-naphthoanthracene, 2,3:6,7-dibenzanthracene, unii-9fqu5ha0uy, 9fqu5ha0uy, pentacen-2-yl, pentacene, 2,7-dibenzanthracene
SLIUAWYAILUBJU-UHFFFAOYSA-N
pentacene
8671
278.34
Fine Crystalline Powder
98%
>300°C
Authentic
Glass bottle
MFCD00003710
15, 7216
Solubility in water: insoluble. Other solubilities: slightly soluble in benzene,soluble in nitrobenzene
C1=CC2=CC3=CC4=CC5=CC=CC=C5C=C4C=C3C=C2C=C1
278.35
CHEBI:33148
98%

Chemical Identifiers

135-48-8
278.35
SLIUAWYAILUBJU-UHFFFAOYSA-N
8671
pentacene
C22H14
MFCD00003710
benzo b naphthacene, lin-dibenzanthracene, lin-naphthoanthracene, 2,3:6,7-dibenzanthracene, unii-9fqu5ha0uy, 9fqu5ha0uy, pentacen-2-yl, pentacene, 2,7-dibenzanthracene
CHEBI:33148
C1=CC2=CC3=CC4=CC5=CC=CC=C5C=C4C=C3C=C2C=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

AIR SENSITIVE

LIGHT SENSITIVE

EINECSNumber : 205-193-7

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only