missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentacene, 98%
CAS: 135-48-8 | C22H14 | 278.35 g/mol
$113.34 - $555.79
Chemical Identifiers
| CAS | 135-48-8 |
|---|---|
| Molecular Formula | C22H14 |
| Molecular Weight (g/mol) | 278.35 |
| MDL Number | MFCD00003710 |
| InChI Key | SLIUAWYAILUBJU-UHFFFAOYSA-N |
| Synonym | benzo b naphthacene, lin-dibenzanthracene, lin-naphthoanthracene, 2,3:6,7-dibenzanthracene, unii-9fqu5ha0uy, 9fqu5ha0uy, pentacen-2-yl, pentacene, 2,7-dibenzanthracene |
| PubChem CID | 8671 |
| ChEBI | CHEBI:33148 |
| IUPAC Name | pentacene |
| SMILES | C1=CC2=CC3=CC4=CC5=CC=CC=C5C=C4C=C3C=C2C=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC129791000
|
Thermo Scientific Chemicals
129791000 |
100 mg | Glass bottle |
Each for $113.34
|
|
||||
|
AC129790010
|
Thermo Scientific Chemicals
129790010 |
1 g | Glass bottle |
Each for $555.79
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 135-48-8 | |
| 278.35 | |
| SLIUAWYAILUBJU-UHFFFAOYSA-N | |
| 8671 | |
| pentacene |
| C22H14 | |
| MFCD00003710 | |
| benzo b naphthacene, lin-dibenzanthracene, lin-naphthoanthracene, 2,3:6,7-dibenzanthracene, unii-9fqu5ha0uy, 9fqu5ha0uy, pentacen-2-yl, pentacene, 2,7-dibenzanthracene | |
| CHEBI:33148 | |
| C1=CC2=CC3=CC4=CC5=CC=CC=C5C=C4C=C3C=C2C=C1 |
Specifications
| 135-48-8 | |
| Black or Blue-Purple | |
| 97.5% min | |
| C22H14 | |
| 100 mg | |
| benzo b naphthacene, lin-dibenzanthracene, lin-naphthoanthracene, 2,3:6,7-dibenzanthracene, unii-9fqu5ha0uy, 9fqu5ha0uy, pentacen-2-yl, pentacene, 2,7-dibenzanthracene | |
| SLIUAWYAILUBJU-UHFFFAOYSA-N | |
| pentacene | |
| 8671 | |
| 278.34 | |
| Fine Crystalline Powder |
| >300°C | |
| Authentic | |
| Glass bottle | |
| MFCD00003710 | |
| 15, 7216 | |
| Solubility in water: insoluble. Other solubilities: slightly soluble in benzene,soluble in nitrobenzene | |
| C1=CC2=CC3=CC4=CC5=CC=CC=C5C=C4C=C3C=C2C=C1 | |
| 278.35 | |
| CHEBI:33148 | |
| 98% | |
| Pentacene, 98% |
Safety and Handling
AIR SENSITIVE
LIGHT SENSITIVE
EINECSNumber : 205-193-7
TSCA : TSCA
RUO – Research Use Only