missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Pentacene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 129791000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Pentacene | |
135-48-8 | |
Black or Blue-Purple | |
97.5% min | |
C22H14 | |
100 mg | |
benzo b naphthacene, lin-dibenzanthracene, lin-naphthoanthracene, 2,3:6,7-dibenzanthracene, unii-9fqu5ha0uy, 9fqu5ha0uy, pentacen-2-yl, pentacene, 2,7-dibenzanthracene | |
SLIUAWYAILUBJU-UHFFFAOYSA-N | |
pentacene | |
8671 | |
278.34 | |
Fine Crystalline Powder |
98% | |
>300°C | |
Authentic | |
Glass bottle | |
MFCD00003710 | |
15, 7216 | |
Solubility in water: insoluble. Other solubilities: slightly soluble in benzene,soluble in nitrobenzene | |
C1=CC2=CC3=CC4=CC5=CC=CC=C5C=C4C=C3C=C2C=C1 | |
278.35 | |
CHEBI:33148 | |
98% |
Chemical Identifiers
135-48-8 | |
278.35 | |
SLIUAWYAILUBJU-UHFFFAOYSA-N | |
8671 | |
pentacene |
C22H14 | |
MFCD00003710 | |
benzo b naphthacene, lin-dibenzanthracene, lin-naphthoanthracene, 2,3:6,7-dibenzanthracene, unii-9fqu5ha0uy, 9fqu5ha0uy, pentacen-2-yl, pentacene, 2,7-dibenzanthracene | |
CHEBI:33148 | |
C1=CC2=CC3=CC4=CC5=CC=CC=C5C=C4C=C3C=C2C=C1 |
Safety and Handling
AIR SENSITIVE
LIGHT SENSITIVE
EINECSNumber : 205-193-7
TSCA : TSCA
RUO – Research Use Only