missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pentafluorophenyl 4-(tetrahydropyran-4-yloxy)benzoate, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC52426DA

 View more versions of this product

Catalog No. CC52426DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Pentafluorophenyl 4-(tetrahydropyran-4-yloxy)benzoate
96.5
97%
C18H13F5O4
1g
BDWCEFHYQPZQRO-UHFFFAOYSA-N
(2,3,4,5,6-pentafluorophenyl) 4-(oxan-4-yloxy)benzoate
24229528
97%
930110-97-7
100.0
Amber Glass Bottle
MFCD09702368
pentafluorophenyl 4-tetrahydropyran-4-yloxy benzoate, pentafluorophenyl 4-oxan-4-yloxy benzoate, pentafluorophenyl 4-oxan-4-yl oxy benzoate, 2,3,4,5,6-pentafluorophenyl 4-oxan-4-yloxy benzoate, 2,3,4,5,6-pentakis fluoranyl phenyl 4-oxan-4-yloxy benzoate, 4-4-oxanyloxy benzoic acid 2,3,4,5,6-pentafluorophenyl ester
FC1=C(F)C(F)=C(OC(=O)C2=CC=C(OC3CCOCC3)C=C2)C(F)=C1F
388.29
388.29

Chemical Identifiers

930110-97-7
388.29
BDWCEFHYQPZQRO-UHFFFAOYSA-N
24229528
FC1=C(F)C(F)=C(OC(=O)C2=CC=C(OC3CCOCC3)C=C2)C(F)=C1F
C18H13F5O4
MFCD09702368
pentafluorophenyl 4-tetrahydropyran-4-yloxy benzoate, pentafluorophenyl 4-oxan-4-yloxy benzoate, pentafluorophenyl 4-oxan-4-yl oxy benzoate, 2,3,4,5,6-pentafluorophenyl 4-oxan-4-yloxy benzoate, 2,3,4,5,6-pentakis fluoranyl phenyl 4-oxan-4-yloxy benzoate, 4-4-oxanyloxy benzoic acid 2,3,4,5,6-pentafluorophenyl ester
(2,3,4,5,6-pentafluorophenyl) 4-(oxan-4-yloxy)benzoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions