missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pentafluorophenyl 4-(tetrahydropyran-4-yloxy)benzoate, 97%, Thermo Scientific™

Chemical Identifiers

CAS 930110-97-7
Molecular Formula C18H13F5O4
Molecular Weight (g/mol) 388.29
MDL Number MFCD09702368
InChI Key BDWCEFHYQPZQRO-UHFFFAOYSA-N
Synonym pentafluorophenyl 4-tetrahydropyran-4-yloxy benzoate, pentafluorophenyl 4-oxan-4-yloxy benzoate, pentafluorophenyl 4-oxan-4-yl oxy benzoate, 2,3,4,5,6-pentafluorophenyl 4-oxan-4-yloxy benzoate, 2,3,4,5,6-pentakis fluoranyl phenyl 4-oxan-4-yloxy benzoate, 4-4-oxanyloxy benzoic acid 2,3,4,5,6-pentafluorophenyl ester
PubChem CID 24229528
IUPAC Name (2,3,4,5,6-pentafluorophenyl) 4-(oxan-4-yloxy)benzoate
SMILES FC1=C(F)C(F)=C(OC(=O)C2=CC=C(OC3CCOCC3)C=C2)C(F)=C1F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC52426CB
SDS View Documents
Thermo Scientific Chemicals
CC52426CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC52426DA
SDS View Documents
Thermo Scientific Chemicals
CC52426DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

930110-97-7
388.29
BDWCEFHYQPZQRO-UHFFFAOYSA-N
24229528
FC1=C(F)C(F)=C(OC(=O)C2=CC=C(OC3CCOCC3)C=C2)C(F)=C1F
C18H13F5O4
MFCD09702368
pentafluorophenyl 4-tetrahydropyran-4-yloxy benzoate, pentafluorophenyl 4-oxan-4-yloxy benzoate, pentafluorophenyl 4-oxan-4-yl oxy benzoate, 2,3,4,5,6-pentafluorophenyl 4-oxan-4-yloxy benzoate, 2,3,4,5,6-pentakis fluoranyl phenyl 4-oxan-4-yloxy benzoate, 4-4-oxanyloxy benzoic acid 2,3,4,5,6-pentafluorophenyl ester
(2,3,4,5,6-pentafluorophenyl) 4-(oxan-4-yloxy)benzoate

Specifications

930110-97-7
100.0
Amber Glass Bottle
MFCD09702368
pentafluorophenyl 4-tetrahydropyran-4-yloxy benzoate, pentafluorophenyl 4-oxan-4-yloxy benzoate, pentafluorophenyl 4-oxan-4-yl oxy benzoate, 2,3,4,5,6-pentafluorophenyl 4-oxan-4-yloxy benzoate, 2,3,4,5,6-pentakis fluoranyl phenyl 4-oxan-4-yloxy benzoate, 4-4-oxanyloxy benzoic acid 2,3,4,5,6-pentafluorophenyl ester
FC1=C(F)C(F)=C(OC(=O)C2=CC=C(OC3CCOCC3)C=C2)C(F)=C1F
388.29
388.29
Pentafluorophenyl 4-(tetrahydropyran-4-yloxy)benzoate
96.5
97%
C18H13F5O4
250mg
BDWCEFHYQPZQRO-UHFFFAOYSA-N
(2,3,4,5,6-pentafluorophenyl) 4-(oxan-4-yloxy)benzoate
24229528
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents