missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Perchloric Acid, ca. 70% Solution in Water, For analysis, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 424030010

 View more versions of this product

Catalog No. AC424030010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Specifications

Perchloric acid
-18.0°C
203.0°C
1ppm max.
0.003% max.
113°C
3.5 mPa.s (20°C)
Solution
MFCD00011325
15, 7267
Solubility in water: soluble.
O[Cl](=O)(=O)=O
100.45
CHEBI:29221
69.0 to 72.0%
ca. 70% Solution in Water
7601-90-3,7732-18-5
1.6600g/mL
Glass bottle
1.66
1 kg
ClHO4
HClO4
01,796; 02,309; 03,220; 06,453; 09,279; 11,402
unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
VLTRZXGMWDSKGL-UHFFFAOYSA-N
perchloric acid
24247
100.46
ACS Reagent

Chemical Identifiers

7601-90-3
100.45
VLTRZXGMWDSKGL-UHFFFAOYSA-N
24247
perchloric acid
ClHO4
MFCD00011325
unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
CHEBI:29221
O[Cl](=O)(=O)=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
May cause fire or explosion; strong oxidizer.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only