missing translation for 'onlineSavingsMsg'
Learn More

Perchloric acid, ACS reagent, ca. 70% solution in water

CAS: 7601-90-3 | ClHO4 | 100.45 g/mol

$158.72 - $378.44

Chemical Identifiers

CAS 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC424032500
SDS View Documents
Thermo Scientific Chemicals
424032500
250 g Glass bottle
Each for $158.72
Only null left
 
AC424030010
SDS View Documents
Thermo Scientific Chemicals
424030010
1 kg Glass bottle
Each for $378.44
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Chemical Identifiers

7601-90-3
100.45
VLTRZXGMWDSKGL-UHFFFAOYSA-N
24247
perchloric acid
ClHO4
MFCD00011325
unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
CHEBI:29221
O[Cl](=O)(=O)=O

Specifications

-18.0°C
203.0°C
1ppm max.
0.003% max.
113°C
3.5 mPa.s (20°C)
Solution
MFCD00011325
15, 7267
Solubility in water: soluble.
O[Cl](=O)(=O)=O
100.45
CHEBI:29221
69.0 to 72.0%
Perchloric acid, ca. 70% Solution in Water
7601-90-3,7732-18-5
1.6600g/mL
Glass bottle
1.66
250 g
ClHO4
HClO4
01,796; 02,309; 03,220; 06,453; 09,279; 11,402
unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
VLTRZXGMWDSKGL-UHFFFAOYSA-N
perchloric acid
24247
100.46
ACS Reagent
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
May cause fire or explosion; strong oxidizer.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only