Perfex™ Floor Brushes
Supplier:  Perfex™ 2524B
 View more versions of this product
Catalog No. 19-042-017A
Explore available promotions
Requires : Requires 19-042-037 or 19-042-040 adjustable polymer handle (sold separately).
Description
Description
Durable polypropylene fibers are fused to a high-impact resistant polymer block, eliminating the problems of fiber fallout and contamination collection at attachment points. A unique socket design instantly locks the brush tightly to a handle. Block and fibers are nonconductive and nonabsorbent, impervious to grease, water, solvents, petroleum products, and sanitizers.
Order Info
Available with a soft-sweeping bristle or stiff-sweeping bristle. Both offer a choice of widths: 18 and 24 in. (46 and 61cm). Come in blue, red, and white to isolate areas of use quickly and visually.
Specifications
Specifications
Blue
24 in.
Stiff-sweeping bristle
Sweeping heavy or coarse material and removing difficult dirt and deposits from unfinished surfaces, wood, concrete, and blacktop
Polypropylene (brush fiber); Polymer brush block