Perfex™ Floor Brushes
$64.52 - $614.22
Products 8
Catalog Number Mfr. No. Style Color Length (English) Price Quantity  
Catalog Number Mfr. No. Style Color Length (English) Price Quantity  
19-042-028A
View Documents
Perfex™
2618B
Soft-sweeping bristle Blue 18 in.
Each for $64.52
Pack of 10 Each for $603.60
19-042-017A
View Documents
Perfex™
2524B
Stiff-sweeping bristle Blue 24 in.
Each for $68.07
Pack of 10 Each for $636.80
19-042-017C
View Documents
Perfex™
2524W
Stiff-sweeping bristle White 24 in.
Each for $68.07
Pack of 10 Each for $636.80
19-042-016B
View Documents
Perfex™
2518R
Stiff-sweeping bristle Red 18 in.
Pack of 10 Each for $614.22
19-042-016C
View Documents
Perfex™
2518W
Stiff-sweeping bristle White 18 in.
Each for $64.83
Pack of 10 Each for $606.50
19-042-016A
View Documents
Perfex™
2518B
Stiff-sweeping bristle Blue 18 in.
Each for $64.83
Pack of 10 Each for $606.50
19-042-017B
View Documents
Perfex™
2524R
Stiff-sweeping bristle Red 24 in.
Each for $68.07
19-042-030A
View Documents
Perfex™
2624B
Soft-sweeping bristle Blue 24 in.
Each for $68.07
Description
Description
Durable polypropylene fibers are fused to a high-impact resistant polymer block, eliminating the problems of fiber fallout and contamination collection at attachment points. A unique socket design instantly locks the brush tightly to a handle. Block and fibers are nonconductive and nonabsorbent, impervious to grease, water, solvents, petroleum products, and sanitizers.
Order Info
Available with a soft-sweeping bristle or stiff-sweeping bristle. Both offer a choice of widths: 18 and 24 in. (46 and 61cm). Come in blue, red, and white to isolate areas of use quickly and visually.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'