Perfex™ Floor Brushes
Supplier:  Perfex™ 2524R
 View more versions of this product
Catalog No. 19-042-017B
Explore available promotions
Requires : Requires 19-042-037 or 19-042-040 adjustable polymer handle (sold separately).
Description
Description
Durable polypropylene fibers are fused to a high-impact resistant polymer block, eliminating the problems of fiber fallout and contamination collection at attachment points. A unique socket design instantly locks the brush tightly to a handle. Block and fibers are nonconductive and nonabsorbent, impervious to grease, water, solvents, petroleum products, and sanitizers.
Order Info
Available with a soft-sweeping bristle or stiff-sweeping bristle. Both offer a choice of widths: 18 and 24 in. (46 and 61cm). Come in blue, red, and white to isolate areas of use quickly and visually.
Specifications
Specifications
Stiff Sweep Floor Brush
For sweeping heavy or coarse material and for removing difficult dirt and deposits from unfinished surfaces, Wood, concrete, blacktop
24 in.
Polypropylene Fiber
Red
Adjustable Polymer Handle
61 cm
Stiff-sweeping bristle