Requires : Requires 19-042-037 or 19-042-040 adjustable polymer handle (sold separately).
Description
Durable polypropylene fibers are fused to a high-impact resistant polymer block, eliminating the problems of fiber fallout and contamination collection at attachment points. A unique socket design instantly locks the brush tightly to a handle. Block and fibers are nonconductive and nonabsorbent, impervious to grease, water, solvents, petroleum products, and sanitizers.
Order Info
Available with a soft-sweeping bristle or stiff-sweeping bristle. Both offer a choice of widths: 18 and 24 in. (46 and 61cm). Come in blue, red, and white to isolate areas of use quickly and visually.
Specifications
Stiff Sweep Floor Brush | |
For sweeping heavy or coarse material and for removing difficult dirt and deposits from unfinished surfaces, Wood, concrete, blacktop | |
24 in. | |
Polypropylene Fiber |
White | |
Adjustable Polymer Handle | |
61 cm | |
Stiff-sweeping bristle |