missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro(2-methyl-2-pentene), 97%

CAS: 1584-03-8 | C6F12 | 300.047 g/mol

$171.23 - $653.16

Identifiants chimiques

CAS 1584-03-8
Molecular Formula C6F12
Molecular Weight (g/mol) 300.047
MDL Number MFCD00015724
InChI Key FAEGGADNHFKDQX-UHFFFAOYSA-N
Synonym perfluoro-2-methyl-2-pentene, perfluoro 2-methylpent-2-ene, hexafluoropropene dimer, perfluoro-2-methylpent-2-ene, 1,1,1,3,4,4,5,5,5-nonafluoro-2-trifluoromethyl pent-2-ene, perfluoro 2-methyl-2-pentene, nonafluoro-2-trifluoromethyl pent-2-ene, 1,1,1,3,4,4,5,5,5-nonafluoro-2-trifluoromethyl pent-4-ene, 2-pentene, 1,1,1,3,4,4,5,5,5-nonafluoro-2-trifluoromethyl, 1,1,1,3,4,4,5,5,5-nonafluoro-2-trifluoromethyl-2-pentene
PubChem CID 74105
IUPAC Name 1,1,1,3,4,4,5,5,5-nonafluoro-2-(trifluoromethyl)pent-2-ene
SMILES C(=C(C(C(F)(F)F)(F)F)F)(C(F)(F)F)C(F)(F)F
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AAA1839406
Afficher les documents
Thermo Scientific Chemicals
A1839406
5 g
chaque for $171.23
Il en reste null
 
AAA1839414
Afficher les documents
Thermo Scientific Chemicals
A1839414
25 g
chaque for $653.16
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

1584-03-8
300.047
FAEGGADNHFKDQX-UHFFFAOYSA-N
74105
C(=C(C(C(F)(F)F)(F)F)F)(C(F)(F)F)C(F)(F)F
C6F12
MFCD00015724
perfluoro-2-methyl-2-pentene, perfluoro 2-methylpent-2-ene, hexafluoropropene dimer, perfluoro-2-methylpent-2-ene, 1,1,1,3,4,4,5,5,5-nonafluoro-2-trifluoromethyl pent-2-ene, perfluoro 2-methyl-2-pentene, nonafluoro-2-trifluoromethyl pent-2-ene, 1,1,1,3,4,4,5,5,5-nonafluoro-2-trifluoromethyl pent-4-ene, 2-pentene, 1,1,1,3,4,4,5,5,5-nonafluoro-2-trifluoromethyl, 1,1,1,3,4,4,5,5,5-nonafluoro-2-trifluoromethyl-2-pentene
1,1,1,3,4,4,5,5,5-nonafluoro-2-(trifluoromethyl)pent-2-ene

Spécifications

1584-03-8
48°C to 54°C
C6F12
MFCD00015724
UN1993
FAEGGADNHFKDQX-UHFFFAOYSA-N
1,1,1,3,4,4,5,5,5-nonafluoro-2-(trifluoromethyl)pent-2-ene
74105
97%
1.62
−12°C (10°F)
ca. 1.3
5 g
perfluoro-2-methyl-2-pentene, perfluoro 2-methylpent-2-ene, hexafluoropropene dimer, perfluoro-2-methylpent-2-ene, 1,1,1,3,4,4,5,5,5-nonafluoro-2-trifluoromethyl pent-2-ene, perfluoro 2-methyl-2-pentene, nonafluoro-2-trifluoromethyl pent-2-ene, 1,1,1,3,4,4,5,5,5-nonafluoro-2-trifluoromethyl pent-4-ene, 2-pentene, 1,1,1,3,4,4,5,5,5-nonafluoro-2-trifluoromethyl, 1,1,1,3,4,4,5,5,5-nonafluoro-2-trifluoromethyl-2-pentene
C(=C(C(C(F)(F)F)(F)F)F)(C(F)(F)F)C(F)(F)F
300.047
300.05
Perfluoro(2-methyl-2-pentene)
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H225
Highly flammable liquid and vapour.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

missing translation for 'einecsNumber' : 216-436-1

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only