missing translation for 'onlineSavingsMsg'
Learn More

Petroleum Ether, B.R. 35 DEG -60°C, ACS, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor P10451LTGL

 View more versions of this product

Catalog No. 18-612-196


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Petroleum Ether
1
20°C to 40°C
Amber Glass Bottle
Reagent
8032-32-4
To Pass Test
1 L
1e-05

Chemical Identifiers

8032-32-4
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.