missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Petroleum Ether, B.R. 35 DEG -60°C, ACS, Spectrum™ Chemical
$193.38 - $1475.65
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18-612-194
|
Spectrum Chemical Mfg Cor
P1045100ML |
100 mL | Amber Glass Bottle |
Each for $193.38
|
|
||||
18-612-195
|
Spectrum Chemical Mfg Cor
P1045500MLGL |
500 mL | Amber Glass Bottle |
Each for $233.26
|
|
||||
18-612-196
|
Spectrum Chemical Mfg Cor
P10451LTGL |
1 L | Amber Glass Bottle |
Each for $302.56
|
|
||||
18-612-197
|
Spectrum Chemical Mfg Cor
P10454LTGL |
4 L | Amber Glass Bottle |
Each for $595.71
|
|
||||
18-612-198
|
Spectrum Chemical Mfg Cor
P104520LTMT |
20 L | Steel Can |
Each for $1,475.65
|
|
||||
Chemical Identifiers
8032-32-4 |
Specifications
8032-32-4 | |
To Pass Test | |
100 mL | |
1e-05 |
1 | |
35°C to 60°C | |
Amber Glass Bottle | |
ACS |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.