Learn More
Petroleum Ether (Certified ACS), Fisher Chemical™
Also meets specifications of the American Oil Chemists Society and the National Cottonseed Producers Association
Supplier: Thermo Fisher Scientific E139S4
Specifications
| Petroleum Ether | |
| Colorless | |
| 38°C | |
| Pass Test | |
| Liquid | |
| 0.630 to 0.660 (at 25°C) | |
| Ligroine, Benzine, Naphtha Petroleum, Naphtha Solvent (Optima/Pesticide/Certified ACS), magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide |
| 8032-32-4 | |
| Pass Test | |
| 4 L | |
| Aluminum Can | |
| 0.001% max. | |
| MFCD00081849 | |
| Certified ACS Reagent |
Chemical Identifiers
| 8032-32-4 | |
| MFCD00081849 | |
| Ligroine, Benzine, Naphtha Petroleum, Naphtha Solvent (Optima/Pesticide/Certified ACS), magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide |
Safety and Handling
DANGER!
Emergency Overview
Flammable liquid and vapor. May cause heritable genetic damage. May cause eye, skin, and respiratory tract irritation. Inhalation may cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.
NFPA
Health:1
Flammability:3
Instability:0
DOTInformation : DOT Class 3, : Flammable Liquid