Learn More


Petroleum Ether (Certified ACS), Fisher Chemical™
Also meets specifications of the American Oil Chemists Society and the National Cottonseed Producers Association
$289.64 - $19978.36
Chemical Identifiers
CAS | 8032-32-4 |
---|---|
MDL Number | MFCD00081849 |
Synonym | Ligroine, Benzine, Naphtha Petroleum, Naphtha Solvent (Optima/Pesticide/Certified ACS), magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
E139-500
|
Thermo Fisher Scientific
E139500 |
500 mL | Glass Bottle |
Each for $289.64
Case of 6 Each for $1,737.84
|
|
|||||
E139-1
|
Thermo Fisher Scientific
E1391 |
1 L | Glass Bottle |
Each for $342.82
Case of 6 Each for $2,056.92
|
|
|||||
E139S-4
|
Fisher Chemical
E139S4 |
4 L | Aluminum Can |
Each for $1,060.27
Case of 4 Each for $4,241.08
|
|
|||||
E139-4
|
Thermo Fisher Scientific
BPE1394 |
4 L | Glass Bottle |
Each for $673.64
Case of 4 Each for $2,694.56
|
|
|||||
E139-20
|
Fisher Chemical
E13920 |
20 L | Steel Pail |
Each for $2,519.18
|
|
|||||
E139-200
|
Thermo Fisher Scientific
E139200 |
200 L | Steel Drum |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | ||||||||||
Chemical Identifiers
8032-32-4 | |
MFCD00081849 | |
Ligroine, Benzine, Naphtha Petroleum, Naphtha Solvent (Optima/Pesticide/Certified ACS), magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide |
Specifications
8032-32-4 | |
Pass Test | |
500 mL | |
Glass Bottle | |
0.001% max. | |
MFCD00081849 | |
Certified ACS Reagent |
Colorless | |
38°C | |
Pass Test | |
Liquid | |
0.630 to 0.660 (at 25°C) | |
Ligroine, Benzine, Naphtha Petroleum, Naphtha Solvent (Optima/Pesticide/Certified ACS), magnesium, bromo 4-methylphenyl, 4-methylphenylmagnesium bromide, bromo 4-methylphenyl magnesium, 4-tolylmagnesium bromide, 4-toluylmagnesium bromide, p-tolyl-magnesium bromide, 4-tolyl magnesium bromide, 4-methylphenylmagnesiumbromide, p-methylphenylmagnesium bromide | |
Petroleum Ether |
Safety and Handling


DANGER!
Emergency Overview
Flammable liquid and vapor. May cause heritable genetic damage. May cause eye, skin, and respiratory tract irritation. Inhalation may cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.
NFPA
Health:1
Flammability:3
Instability:0
DOTInformation : DOT Class 3, : Flammable Liquid