missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenazine methosulfate, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0210095510

 View more versions of this product

Catalog No. ICN10095510


Explore available promotions

Description

Description

Specifications

Specifications

Phenazine methosulfate
Yellow
10 g
RXGJTUSBYWCRBK-UHFFFAOYSA-M
5-methylphenazin-5-ium;methyl sulfate
9285
306.3
299-11-6
C14H14N2O4S
phenazine methosulfate, 5-methylphenazin-5-ium methyl sulfate, 5-methylphenazinium methyl sulfate, n-methylphenazonium methosulfate, phenazine methosulphate, methylphenazonium methosulfate, 5-methylphenazine methylsulfate, n-methylphenazinium methosulfate, n-methylphenazonium methosulphate, phenazinium, 5-methyl-, methyl sulfate
C[N+]1=C2C=CC=CC2=NC3=CC=CC=C31.COS(=O)(=O)[O-]
306.336
CHEBI:8055
Crystals

Chemical Identifiers

299-11-6
306.336
phenazine methosulfate, 5-methylphenazin-5-ium methyl sulfate, 5-methylphenazinium methyl sulfate, n-methylphenazonium methosulfate, phenazine methosulphate, methylphenazonium methosulfate, 5-methylphenazine methylsulfate, n-methylphenazinium methosulfate, n-methylphenazonium methosulphate, phenazinium, 5-methyl-, methyl sulfate
CHEBI:8055
C[N+]1=C2C=CC=CC2=NC3=CC=CC=C31.COS(=O)(=O)[O-]
C14H14N2O4S
RXGJTUSBYWCRBK-UHFFFAOYSA-M
9285
5-methylphenazin-5-ium;methyl sulfate
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at 2° to 8°C.

SDS
Documents

Documents

Promotions

Promotions