Learn More
Phenethyl alcohol, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 130180050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Phenethyl alcohol | |
-27.0°C | |
1.0200g/mL | |
102°C | |
98.5% min. (GC) | |
C8H10O | |
C6H5CH2CH2OH | |
5 mL | |
1.02 | |
phenethyl alcohol, benzeneethanol, phenylethyl alcohol, benzyl carbinol, phenylethanol, phenethanol, 2-phenylethyl alcohol, 2-phenyl-ethanol, beta-phenylethanol, 2-phenylethan-1-ol | |
WRMNZCZEMHIOCP-UHFFFAOYSA-N | |
2-phenylethanol | |
6054 | |
CHEBI:49000 | |
99% |
60-12-8 | |
Colorless | |
219.0°C | |
Authentic | |
Glass bottle | |
1.5300 to 1.5350 | |
MFCD00002886 | |
06, 478 | |
15, 7338 | |
Solubility in water: 20g/L (20°C). Other solubilities: miscible with alcohol and ether | |
OCCC1=CC=CC=C1 | |
122.17 | |
7.58 mPa.s (25°C) | |
122.17 | |
Liquid |
Chemical Identifiers
60-12-8 | |
122.17 | |
WRMNZCZEMHIOCP-UHFFFAOYSA-N | |
6054 | |
OCCC1=CC=CC=C1 |
C8H10O | |
MFCD00002886 | |
phenethyl alcohol, benzeneethanol, phenylethyl alcohol, benzyl carbinol, phenylethanol, phenethanol, 2-phenylethyl alcohol, 2-phenyl-ethanol, beta-phenylethanol, 2-phenylethan-1-ol | |
CHEBI:49000 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED
GHS Signal Word: Warning
EINECSNumber : 200-456-2
RUO – Research Use Only