Learn More
Phenethyl alcohol, 99%, Thermo Scientific Chemicals
$36.56 - $473.05
Chemical Identifiers
CAS | 60-12-8 |
---|---|
Molecular Formula | C8H10O |
Molecular Weight (g/mol) | 122.17 |
MDL Number | MFCD00002886 |
InChI Key | WRMNZCZEMHIOCP-UHFFFAOYSA-N |
Synonym | phenethyl alcohol, benzeneethanol, phenylethyl alcohol, benzyl carbinol, phenylethanol, phenethanol, 2-phenylethyl alcohol, 2-phenyl-ethanol, beta-phenylethanol, 2-phenylethan-1-ol |
PubChem CID | 6054 |
ChEBI | CHEBI:49000 |
SMILES | OCCC1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC130180050
|
Thermo Scientific Chemicals
130180050 |
5 mL | Glass bottle |
Each for $36.56
|
|
||||
AC130181000
|
Thermo Scientific Chemicals
130181000 |
100 mL | Glass bottle |
Each for $72.42
|
|
||||
AC130182500
|
Thermo Scientific Chemicals
130182500 |
250 mL | Glass bottle |
Each for $111.32
|
|
||||
AC130180010
|
Thermo Scientific Chemicals
130180010 |
1 L | Glass bottle |
Each for $209.14
Case of 4 Each for $0.00
|
|
||||
AC130180025
|
Thermo Scientific Chemicals
130180025 |
2.5 L | Glass bottle |
Each for $473.05
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
60-12-8 | |
122.17 | |
WRMNZCZEMHIOCP-UHFFFAOYSA-N | |
6054 | |
OCCC1=CC=CC=C1 |
C8H10O | |
MFCD00002886 | |
phenethyl alcohol, benzeneethanol, phenylethyl alcohol, benzyl carbinol, phenylethanol, phenethanol, 2-phenylethyl alcohol, 2-phenyl-ethanol, beta-phenylethanol, 2-phenylethan-1-ol | |
CHEBI:49000 |
Specifications
60-12-8 | |
Colorless | |
219.0°C | |
Authentic | |
Glass bottle | |
1.5300 to 1.5350 | |
MFCD00002886 | |
06, 478 | |
15, 7338 | |
Solubility in water: 20g/L (20°C). Other solubilities: miscible with alcohol and ether | |
OCCC1=CC=CC=C1 | |
122.17 | |
7.58 mPa.s (25°C) | |
122.17 | |
Liquid |
-27.0°C | |
1.0200g/mL | |
102°C | |
98.5% min. (GC) | |
C8H10O | |
C6H5CH2CH2OH | |
5 mL | |
1.02 | |
phenethyl alcohol, benzeneethanol, phenylethyl alcohol, benzyl carbinol, phenylethanol, phenethanol, 2-phenylethyl alcohol, 2-phenyl-ethanol, beta-phenylethanol, 2-phenylethan-1-ol | |
WRMNZCZEMHIOCP-UHFFFAOYSA-N | |
2-phenylethanol | |
6054 | |
CHEBI:49000 | |
99% | |
Phenethyl alcohol |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED
GHS Signal Word: Warning
EINECSNumber : 200-456-2
RUO – Research Use Only