missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phenol Standard, 1000 ppm C6H5OH (0.100% (w/w)), Ricca Chemical
Supplier: Ricca Chemical Company 574032
Specifications
Phenol Standard | |
Amber Glass Bottle | |
Colorless | |
C6H6O | |
ISWSIDIOOBJBQZ-UHFFFAOYSA-N | |
phenol | |
996 | |
1 mL = 1 mg Phenol |
108-95-2,7732-18-5 | |
1 L | |
0.100% (w/w) C6H5OH | |
MFCD00002143 | |
OC1=CC=CC=C1 | |
94.11 | |
CHEBI:15882 | |
Liquid |
Chemical Identifiers
108-95-2 | |
94.11 | |
ISWSIDIOOBJBQZ-UHFFFAOYSA-N | |
CHEBI:15882 | |
OC1=CC=CC=C1 |
C6H6O | |
MFCD00002143 | |
996 | |
phenol |