missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenol Standard, 1000 ppm C6H5OH (0.100% (w/w)), Ricca Chemical

$91.82 - $297.16

Chemical Identifiers

CAS 108-95-2
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
PubChem CID 996
ChEBI CHEBI:15882
IUPAC Name phenol
SMILES OC1=CC=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
5740-4
SDS View Documents
Ricca Chemical Company
RI57404
120 mL Amber Glass Bottle
Each for $91.82
 
5740-16
View Documents
Ricca Chemical Company
574016
500 mL Amber Glass Bottle
Each for $194.88
 
5740-32
View Documents
Ricca Chemical Company
574032
1 L Amber Glass Bottle
Each for $297.16
 
Description

Description

Specifications

Chemical Identifiers

108-95-2
94.11
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
CHEBI:15882
OC1=CC=CC=C1
C6H6O
MFCD00002143
996
phenol

Specifications

108-95-2,7732-18-5
120 mL
0.100% (w/w) C6H5OH
MFCD00002143
OC1=CC=CC=C1
94.11
CHEBI:15882
Liquid
Amber Glass Bottle
Colorless
C6H6O
ISWSIDIOOBJBQZ-UHFFFAOYSA-N
phenol
996
1 mL = 1 mg Phenol
Phenol Standard
SDS
missing translation for 'documents'

missing translation for 'documents'