Fisherbrand™ Phenolic Tall Cabinet
Fisherbrand™ Phenolic Casework is designed to rugged use in wet areas where corrosion or rust are of concern.
 View more versions of this product
Catalog No. FBPH30336P
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Rust, rot, mold resistant construction.
  • Cabinets will not absorb odors, are easy to clean and are virtually maintenance free.
  • Standard cabinet construction is from 0.5 in. thick chemically resistant phenolic panels in a full overlay style.
  • All colors have a solid black core
Specifications
Specifications
36 in.
36 x 22 x 84.6 in.
84.6 in.
Solid Door
Tall
2
Phenolic
2 years
214.9 cm
22 in.
55.8 cm
91.4 cm
Platinum