missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenyl p-tolyl sulfone, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0312818

 View more versions of this product

Catalog No. AAL0312818


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Phenyl p-tolyl sulfone
125°C to 129°C
C13H12O2S
50 g
phenyl p-tolyl sulfone, sulfone, phenyl p-tolyl, 4-methylphenyl phenyl sulfone, benzene, 1-methyl-4-phenylsulfonyl, 1-methyl-4-phenylsulfonyl benzene, 4-methyldiphenylsulfone, p-tolyl phenyl sulfone, p-phenylsulphonyl toluene, 1-benzenesulfonyl-4-methylbenzene, 1-benzenesulfonyl-4-methyl-benzene
CC1=CC=C(C=C1)S(=O)(=O)C2=CC=CC=C2
232.297
232.3
640-57-3
Odorless
MFCD00025038
1872872
YBRXHRWLEFCFEG-UHFFFAOYSA-N
1-(benzenesulfonyl)-4-methylbenzene
69497
≥98%

Chemical Identifiers

640-57-3
232.297
YBRXHRWLEFCFEG-UHFFFAOYSA-N
69497
CC1=CC=C(C=C1)S(=O)(=O)C2=CC=CC=C2
C13H12O2S
MFCD00025038
phenyl p-tolyl sulfone, sulfone, phenyl p-tolyl, 4-methylphenyl phenyl sulfone, benzene, 1-methyl-4-phenylsulfonyl, 1-methyl-4-phenylsulfonyl benzene, 4-methyldiphenylsulfone, p-tolyl phenyl sulfone, p-phenylsulphonyl toluene, 1-benzenesulfonyl-4-methylbenzene, 1-benzenesulfonyl-4-methyl-benzene
1-(benzenesulfonyl)-4-methylbenzene
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 211-364-7

RTECSNumber : WR6880000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only